Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C14orf93 Mouse anti-Human, Mouse, Clone: 2-F6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275425
Description
C14orf93 Monoclonal antibody specifically detects C14orf93 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
C14orf93 | |
2-F6 | |
Unconjugated | |
TBS (pH7.4), 0.5%BSA, 50%Glycerol with 0.05% Sodium Azide | |
chromosome 14 open reading frame 93, EC 2.4.2.18, EC 2.4.2.21, FLJ12154, hypothetical protein LOC60686 | |
Mouse | |
IgG1 | |
Protein A purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Purified | |
C14orf93 | |
Recombinant protein | |
57 kDa | |
100 μL | |
Primary | |
60686.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title