Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C15orf2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550617
Description
The antibody detects endogenous levels of total C15orf2 / NPAP1 protein.
May be involved in spermatogenesis.
Specifications
C15orf2 | |
Polyclonal | |
Unconjugated | |
NPAP1 | |
C15orf2; NPAP1; nuclear pore associated protein 1; nuclear pore-associated protein 1; protein C15orf2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23742 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NZP6 | |
NPAP1 | |
Synthetic peptide corresponding to a region derived from internal residues of human nuclear pore associated protein 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction