Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C15orf23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559828
Description
Immunogen sequence: TVYSLQPPSA LSGGQPADTQ TRATSKSLLP VRSKEVDVSK QLHSGGPEND VTKITKLRRE NGQMKATDTA TRRNVRKGYK PLSKQ Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 64%.
C15orf23 is an essential component of the mitotic spindle required for faithful chromosome segregation and progression into anaphase. It promotes the metaphase-to-anaphase transition and is required for chromosome alignment, normal timing of sister chromatid segregation, and maintenance of spindle pole architecture. The astrin (SPAG5)-kinastrin (SKAP) complex promotes stable microtubule-kinetochore attachments.
Specifications
C15orf23 | |
Polyclonal | |
Unconjugated | |
KNSTRN | |
1700025D04Rik; C10H15orf23; C15orf23; D2Ertd750e; HSD11; Kinastrin; kinetochore localized astrin (SPAG5) binding protein; kinetochore localized astrin/SPAG5 binding protein; kinetochore-localized astrin/SPAG5 binding; kinetochore-localized astrin/SPAG5 binding protein; kinetochore-localized astrin/SPAG5-binding protein; kinetochore-localized astrin-binding protein; Knstrn; putative TRAF4-associated factor 1; SKAP; small kinetochore associated protein; small kinetochore-associated protein; small kinetochore-associated protein, kinetochore-localized astrin-binding protein, TRAF4 associated factor 1; TRAF4 associated factor 1; TRAF4 associated factor 1 homolog; TRAF4AF1; TRAF4-associated factor 1; Unknown (protein for MGC:134039) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90417 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y448 | |
KNSTRN | |
Recombinant protein corresponding to Human C15orf23. Recombinant protein control fragment (Product #RP-98059). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction