Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C15orf27 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA5107294

Catalog No. PIPA5107294


Add to cart

Description

Description

Sequence homology: Cow: 93%; Dog: 79%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 86%; Rat: 100%

C15orf27 is a voltage-sensor protein present on the post-synaptic side of glutamatergic mossy fibers and granule cells in the cerebellum. Despite the presence of a voltage-sensor segment, it does not form a functional ion channel and its precise role remains unclear. The protein undergoes both rapid and slow structural rearrangements in response to changes in voltage, and contains a zinc-binding site that can regulate the slow conformational transition.
Specifications

Specifications

C15orf27
Polyclonal
Unconjugated
TMEM266
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
123591
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Q2M3C6
TMEM266
Synthetic peptide located within the following region: PAGSAQTSPE LEHRVSLFNQ KNQEGFTVFQ IRPVIHFQPT VPMLEDKFRS
Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit