Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C15orf40 Mouse anti-Human, Clone: OTI1D9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525172

Catalog No. PIMA525172


Add to cart

Description

Description

C15orf40 is a protein coding gene.
Specifications

Specifications

C15orf40
Monoclonal
0.52 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WUR7
UPF0235 protein C15orf40, C15orf40
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
123207
Western Blot
OTI1D9
Unconjugated
C15ORF40
Liquid
C15ORF40
Full length human recombit protein of C15orf40 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit