Learn More
Alfa Aesar™ C16 Ceramide, 98%
Manufacturer:  Alfa Aesar™ J67093MA
 View more versions of this product
Catalog No. AAJ67093MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
10mg
Crystalline solid
An endogenous ceramide that acts as a lipid second messenger to regulate apoptosis and stress signaling.
24696-26-2
537.91
Soluble in DMF at 0.15mg/ml
-30°C to -10°C
C16 Ceramide, 98%
98%
C34H67NO3
D-erythro-N-Palmitoylsphingosine; N-Hexadecanoyl-D-erythro-sphingosine