Learn More
Cayman ChemicalSupplier Diversity Partner C16 Sphingomyelin (d18:1/16:0), 6254-89-3, 1 mg DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 100079461
A form of sphingomyelin containing palmitate (16:0), the most common form of sphingomeylin found in eggs; interacts with cholesterol in lipid rafts; produces C16 ceramide when processed by sphingomyelinases
Catalog No. NC1472670