Learn More
Aviva Systems Biology Corp C16orf46 Antibody - N-terminal region (ARP52898_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52898_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C16orf46. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C16orf46. Alternate names: FLJ32702ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60863