Learn More
Aviva Systems Biology Corp C16orf48 Antibody - middle region (ARP51295_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51295_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C16orf48. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C16orf48. Alternate names: DAKV6410, DKFZP434A1319, C16orf48ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60539