Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C16orf48 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113358
Description
C16orf48 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The exact function of C16orf48 remains unknown.
Specifications
C16orf48 | |
Polyclonal | |
Unconjugated | |
ENKD1 | |
AI606951; C16orf48; C18H16orf48; cp048; DAKV6410; E130303B06Rik; ENKD1; enkurin domain containing 1; enkurin domain-containing protein 1; hypothetical protein LOC524652; RGD1307357; uncharacterized protein C16orf48 homolog; UNQ6410/PRO21183 | |
Rabbit | |
Protein G | |
RUO | |
102124, 84080 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q7TSV9, Q9H0I2 | |
ENKD1 | |
Recombinant Human Enkurin domain-containing protein 1 protein (1-346AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction