Learn More
C16orf70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 206021AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-2983
Add to Cart
Description
Description
The MW of this protein is 48 kDa, and our antibody specifically recognizes the 48 kDa protein.
The function of C20orf30 has not been widely studied, and is yet to be fully elucidated.
Specifications
Specifications
C16orf70
Polyclonal
Unconjugated
C16orf70
C16orf6, C16orf70, lin 10, LIN10, UPF0183 protein C16orf70
Rabbit
Antigen Affinity Chromatography
RUO
102547323, 80262
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BSU1
C16orf70, LOC102547323
C16orf70 Fusion Protein Ag14576
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions