Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C16orf71 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA561981

Catalog No. PIPA561981


Only null left
Add to Cart

Description

Description

Immunogen sequence: MASNDKGMAP SLGSPWASQM GPWDAILKAV KDQLPSLDSD SPLSDYGEEE LFIFQRNQTS LIPDLSEELA EDPADG Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 63%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

C16orf71
Polyclonal
Unconjugated
C16orf71
C16orf71; chromosome 16 open reading frame 71; uncharacterized protein C16orf71
Rabbit
Antigen Affinity Chromatography
RUO
146562
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IYS4
C16orf71
Recombinant protein corresponding to Human C16orf71. Recombinant protein control fragment (Product #RP-104277).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.