Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C16orf71 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549317
Description
Specifications
C16orf71 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8IYS4 | |
C16ORF71 | |
KLH conjugated synthetic peptide between 320-348 amino acids from the Central region of human C16orf71. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
C16orf71 | |
Uncharacterized protein C16orf71, C16orf71 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
146562 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction