Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C16orf90 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571062
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 100%.
C16orf90 is a protein coding gene.
Specifications
C16orf90 | |
Polyclonal | |
Unconjugated | |
C16orf90 | |
C16orf90; chromosome 16 open reading frame 90; uncharacterized protein C16orf90 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
646174 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A8MZG2 | |
C16orf90 | |
synthetic peptide directed towards the N-terminal region of human C16orf90. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction