Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C16orf90 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | C16orf90 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB124283
|
Novus Biologicals
NBP309691100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
C16orf90 Polyclonal specifically detects C16orf90 in Human samples. It is validated for Western Blot.Specifications
C16orf90 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
646174 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human C16orf90 (NP_001073993). Peptide sequence APPNIYEGGLGSPQPQCPSAQGSKPKNFRLRHLRGLGLYLESHPPPTGQC | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title