Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C17orf28 Mouse anti-Canine, Human, Rat, Clone: OTI2F4, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA501311
Description
CQ028 may play an important role in the development of cancers in a broad range of tissues.Specifications
C17orf28 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8IV36 | |
17orf28, C17orf28, DMC1, HID-1 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
283987, 287822, 483305 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2F4 | |
Unconjugated | |
HID1 | |
Liquid | |
HID1 | |
Full length human recombit protein of human C17orf28 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Canine, Human, Rat |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title