Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf37 Mouse anti-Human, Clone: OTI1E8, liquid, TrueMAB™
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA504600

Encompass_Preferred

Catalog No. 50-167-5253


Add to Cart

Description

Description

During the past decade, the role of the ERBB2(neu/HER2) oncogene as an important predictor of patient outcome and response to various therapies in breast cancer has been clearly established. The C35 (C17orf37) is a novel tumor biomarker abundantly expressed in breast cancer. Identification of shared tumor-specific targets is useful in developing broadly applicable therapies. The C35 gene is located on chromosome 17q12, 505nucleotides from the 3′ end of the ERBB2oncogene, the antigenic target for trastuzumab(Herceptin) therapy. The chromosomal arrangement of the genes encoding C35 andERBB2 is tail to tail. The ORF 37 open reading frame encodes a 12-kDa protein of unknown function.
Specifications

Specifications

C17orf37
Monoclonal
0.94 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BRT3
MIEN1
Full length human recombit protein of human C17orf37 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Western Blot
OTI1E8
Unconjugated
MIEN1
C17orf37, C35, ORB3, RDX12, XTP4
Mouse
Affinity Chromatography
RUO
84299
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit