Learn More
C17orf46 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 246291AP
Catalog No. 50-173-4462
Add to cart
Description
Description
C17orf46, also named as SPATA32 and TEX34, mainly expressed in Testis. It interacts with syntaxin-1 and ACTB. It ft.s about 42-52 kDa in our detection.
Specifications
Specifications
C17orf46
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96LK8
C17orf46
Rabbit
IgG
150 μL
-20°C
Primary
124783
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SPATA32
Liquid
SPATA32
C17orf46 Fusion Protein Ag18239
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human