Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf53 Mouse anti-Human, DyLight 650, Clone: 5F3H1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237406C

Catalog No. NBP237406C

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C17orf53
5F3H1
Western Blot, ELISA
Purified
C17orf53
Purified recombinant fragment of human C17ORF53 (AA: 282-527) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
chromosome 17 open reading frame 53, FLJ11594, hypothetical protein LOC78995, MGC3130
Mouse
IgG2a
0.1 ml
Primary
78995
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit