Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C17orf74 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554860
Description
Immunogen sequence: LPPPSLYLSP ELRCMPKRVE ARSELRLQSY GRHGSQSRLW GNVEAEQWAS SPPPPHRLPP NPSWVPVGHS PYPSVGWMLY DSWDQ Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
C17orf74 is a protein coding gene.
Specifications
C17orf74 | |
Polyclonal | |
Unconjugated | |
SPEM2 | |
C17orf74; chromosome 17 open reading frame 74; SPEM2; uncharacterized protein C17orf74; Uncharacterized protein SPEM2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201243 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0P670 | |
SPEM2 | |
Recombinant protein corresponding to Human C17orf74. Recombinant protein control fragment (Product #RP-93505). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction