Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf80 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549463

Catalog No. PA549463

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C17orf80
Polyclonal
PBS with 0.09% sodium azide
Q9BSJ5
Uncharacterized protein C17orf80, Cell migration-inducing gene 3 protein, Human lung cancer oncogene 8 protein, HLC-8, C17orf80
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55028
Western Blot
Unconjugated
C17orf80
Liquid
C17ORF80
KLH conjugated synthetic peptide between 60-93 amino acids from human C17orf80.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit