Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C17orf80 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | C17orf80 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159973
|
Novus Biologicals
NBP159973 |
100 μL |
Each of 1 for $436.00
|
|
Description
C17orf80 Polyclonal specifically detects C17orf80 in Human samples. It is validated for Western Blot.Specifications
C17orf80 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Cell migration-inducing gene 3 protein, chromosome 17 open reading frame 80, FLJ20721, HLC-8, HLC-8migration-inducing protein 3, Human lung cancer oncogene 8 protein, lung cancer-related protein 8, MIG3, SPEP1 | |
C17ORF80 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q9BSJ5 | |
55028 | |
Synthetic peptides corresponding to C17ORF80 The peptide sequence was selected from the N terminal of C17ORF80. Peptide sequence MSDNPPRMEVCPYCKKPFKRLKSHLPYCKMIGPTIPTDQKVYQSKPATLP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title