Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C17orf99 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571295
Description
This target displays homology in the following species: Dog: 85%; Human: 100%; Rat: 77%.
Probable B cell-associated cytokine that plays a role in the regulation of humoral immune responses. Involved in lymphocyte B cell development and immunoglobulin/IgA production.
Specifications
C17orf99 | |
Polyclonal | |
Unconjugated | |
C17orf99 | |
C17orf99; chromosome 17 open reading frame 99; GLPG464; IL40; IL-40; Interleukin-40; Protein IL-40; uncharacterized protein C17orf99; UNQ464; UNQ464/PRO809 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100141515 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6UX52 | |
C17orf99 | |
synthetic peptide directed towards the N-terminal region of human C17orf99. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction