Learn More
CERILLIANT™ Crotonyl fentanyl Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco C1870.5ML
Catalog No. 01-001-134
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
A certified Snap-N-Spike™ solution of crotonyl fentanyl suitable for use as starting material in calibrators and controls for LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications.
Specifications
Specifications
67-56-1
9.7°C (49.5°F) - closed cup
C23H28N2O · HCl
0.5 mL
100 μg/mL in methanol (as free base)
384.94
Ampule
-20°C
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation