Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C18orf32 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559354
Description
Immunogen sequence: PFVSRIWPKK AIQESNDTNK GKVNFKGADM NGLPTKGPTE ICDK Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 64%.
May activate the NF-kappa-B signaling pathway. [UniProt].
Specifications
C18orf32 | |
Polyclonal | |
Unconjugated | |
C18orf32 | |
C18orf32; chromosome 18 open reading frame 32; putative NF-kappa-B-activating protein 200; putative NFkB activating protein; UPF0729 protein C18orf32 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
497661 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TCD1 | |
C18orf32 | |
Recombinant protein corresponding to Human C18orf32. Recombinant protein control fragment (Product #RP-98261). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction