Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C18orf32 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544520
Description
Peptide sequence: PLVSPFVSRI WPKKAIQESN DTNKGKVNFK GADMNGLPTK GPTEICDKKK Sequence homology: Cow: 77%; Human: 100%.
May activate the NF-kappa-B signaling pathway. [UniProt].
Specifications
C18orf32 | |
Polyclonal | |
Unconjugated | |
C18orf32 | |
C18orf32; chromosome 18 open reading frame 32; putative NF-kappa-B-activating protein 200; putative NFkB activating protein; UPF0729 protein C18orf32 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
497661 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TCD1 | |
C18orf32 | |
Synthetic peptide directed towards the middle region of human C18orf32. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction