Learn More
Aviva Systems Biology Corp C18orf54 Antibody - middle region (ARP53273_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53273_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C18orf54. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C18orf54. Alternate names: MGC33382, LAS2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-60987