Learn More
Aviva Systems Biology Corp C19orf15 Antibody - C-terminal region (ARP49638_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49638_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C19orf15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C19orf15. Alternate names: DKFZP434A1022, FLJ38899, FLJ46353, MGC125664, C19orf15ApplicationWB. Species reactivity: Guinea Pig, Human, Rat
Catalog No. 50-014-60132