Learn More
C19orf34 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 243531AP
Catalog No. 50-173-4313
Add to cart
Specifications
Specifications
C19orf34
0.24 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8NCQ2
C19orf34
Rabbit
IgG
150 μL
-20°C
Primary
255193
Western Blot
Unconjugated
CSNK1G2-AS1
Liquid
CSNK1G2-AS1
C19orf34 Fusion Protein Ag19528
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human