Learn More
C19orf43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 194201AP
Catalog No. 50-173-2395
Add to cart
Description
Description
The antibody was raised from the C19orf43 protein. A band at ~28kd was detected in Western Blot with human placenta, 293 cell, and A431 cell lysates.
C19orf43, also named as MGC2803 and fSAP18, is a protein coding gene. Exoribonuclease that is part of the telomerase RNA 3 ft. end processing complex and which has the ability to all four unpaired RNA nucleotides from 5 ft. end or 3 ft. end with higher efficiency for purine bases.
Specifications
Specifications
C19orf43
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BQ61
C19orf43, fSAP18
Rabbit
IgG
150 μL
-20°C
Primary
288920, 68544, 79002
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
C19orf43
Liquid
C19orf43, Trir
C19orf43 Fusion Protein Ag13747
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat