Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf45 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19147120UL

 View more versions of this product

Catalog No. NBP1914720


Add to cart

Description

Description

C19orf45 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

C19orf45
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
C19ORF45
Synthetic peptide directed towards the middle region of human C19orf45. Peptide sequence QALPGPPALRCKRASSGVELGDCKISYGSTCSEQKQAYRPQDLPEDRYDK.
56 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:1000
NP_940936
chromosome 19 open reading frame 45, FLJ35784, FLJ56642, hypothetical protein LOC374877
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
374877
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit