Learn More
C19orf61 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 247971AP
Catalog No. 50-173-4549
Add to cart
Description
Description
This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins.
Specifications
Specifications
C19orf61
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H0W8
C19orf61, F17127_1, Protein smg 9 homolog, Protein SMG9, SMG9
Rabbit
IgG
150 μL
-20°C
Primary
56006
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
SMG9
Liquid
SMG9
C19orf61 Fusion Protein Ag14965
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human