Learn More
C19ORF80 Mouse anti-Human, Clone: OTI4F1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF807377
Catalog No. 50-166-8243
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Betatrophin protein, also known as the hepatocellular carcinoma-associated protein TD26, is an adipocyte- and liver-enriched insulin targeted gene that is thought to play a role in lipid metabolism be regulating serum triglyceride levels. Betatrophin is closely related to the angiopoietin-like (ANGPTL) protein family. Betatrophin mRNA increases in the cold environment and after treatment with a high-fat diet, while its mRNA levels decrease in response to fasting, suggesting that it may play a role in mediating thermogenesis and energy homeostasis.
Specifications
Specifications
C19ORF80
Monoclonal
Unconjugated
ANGPTL8
Lyophilized
ANGPTL8
Human recombit protein fragment corresponding to amino acids 22-198 of human C19ORF80 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI4F1
PBS with 8% trehalose and no preservative; pH 7.3
Q6UXH0
C19orf80, PRO1185, PVPA599, RIFL, TD26
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
55908