Learn More
Aviva Systems Biology Corp C1D Antibody - middle region (ARP50534_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50534_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1D. Alternate names: MGC12261, MGC14659, SUNCOR, LRP1, hC1D, SUN-CoRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60369