Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1GalT1 Mouse anti-Human, Clone: 5B6, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $368.50

Specifications

Antigen C1GalT1
Clone 5B6
Applications Immunocytochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABT1555100
View Documents
MilliporeSigma
MABT1555100UG
100 μg
Each for $368.50
Add to cart
 
MABT155525
View Documents
MilliporeSigma
MABT155525UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

C1GalT1 Monoclonal antibody specifically detects C1GalT1 in Human samples. It is validated for Immunocytochemistry.
Specifications

Specifications

C1GalT1
Immunocytochemistry
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Monoclonal
Protein G purified
5B6
Unconjugated
Mouse
Human
Recombinant fragment corresponding to the Lumenal region of human C1GalT1.
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit