Learn More
C1orf104 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 243601AP
Catalog No. 50-173-4317
Add to cart
Description
Description
RUSC1-AS1 is a protein coding gene.
Specifications
Specifications
C1orf104
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q66K80
C1orf104, RP11 21N7.3, RUSC1 antisense gene protein 1, RUSC1 antisense RNA 1, RUSC1-AS1
Rabbit
IgG
150 μL
-20°C
Primary
284618
Western Blot
Unconjugated
RUSC1-AS1
Liquid
RUSC1-AS1
C1orf104 Fusion Protein Ag19536
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human