Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf112 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555082
Description
Immunogen sequence: FANSLLHYAK EFLPFLSDSC CTLHQLYLQI HSKFPPSLYA TRISKAHQEE IAGAFLVTLD PLISQLLTFQ PFMQVVLDSK LDLPCELQFP QCLLLVVVMD KLPSQPKEVQ TL Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.

Specifications
C1orf112 | |
Polyclonal | |
Unconjugated | |
C1orf112 | |
C1orf112; chromosome 1 open reading frame 112; uncharacterized protein C1orf112 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55732 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NSG2 | |
C1orf112 | |
Recombinant protein corresponding to Human C1orf112. Recombinant protein control fragment (Product #RP-93973). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction