Learn More
C1orf116 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148881AP
Catalog No. 50-172-9808
Add to cart
Description
Description
Two isoforms of SARG are produced by alternative splicing encoding 64 kDa and 38 kDa proteins, and both isoforms can be recognized by this antibody (Product No. 14888-1-AP).
Specifically androgen-regulated gene protein (SARG), encoded by C1orf116, is a 601-aa protein considered putative androgen-specific receptor. SARG mRNA is highly expressed in prostate and can be up-regulated by androgens, but not by glucocorticoids. Little is known about the function of SARG to date.
Specifications
Specifications
C1orf116
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BW04
C1orf116, SARG
Rabbit
IgG
150 μL
-20°C
Primary
212439, 79098
Western Blot
Unconjugated
SARG
Liquid
AA986860, SARG
C1orf116 Fusion Protein Ag6675
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse