Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

C1orf159/RIVIG Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19130720UL

 View more versions of this product

Catalog No. NBP19130720



Description

Description

C1orf159/RIVIG Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

C1orf159/RIVIG
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
C1ORF159
Synthetic peptide directed towards the middle region of human C1orf159. Peptide sequence GQSQGALWVCPQTGLPGSGSRPPLPGSPGDPPTRQGQGRIWLVPPALDLS.
Immunogen affinity purified
RUO
Primary
54991
Western Blot
Western Blot 0.2-1 ug/ml
NP_060361
chromosome 1 open reading frame 159, RP11-465B22.4
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit