Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf168 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546346
Description
Peptide sequence: KFPAGVSPKG DIGGTQSTQI LANGKPLSSN HKQRTPYCSS SESQPLQPQK Sequence homology: Human: 100%.
Adapter protein that plays a role in T-cell receptor (TCR)-mediated activation of signaling pathways. Required for T-cell activation and integrin-mediated T-cell adhesion in response to TCR stimulation (PubMed:27335501). [UniProt].
Specifications
C1orf168 | |
Polyclonal | |
Unconjugated | |
FYB2 | |
Activation-dependent, raft-recruited ADAP-like phosphoprotein; ARAP; C1orf168; chromosome 1 open reading frame 168; FYB2; FYN-binding protein 2; RGD1564074; RP4-758N20.2; similar to novel protein; uncharacterized protein C1orf168 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
199920 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VWT5 | |
FYB2 | |
Synthetic peptide directed towards the N-terminal of human C1orf168 (aa 29-78). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction