Learn More
Aviva Systems Biology Corp C1orf184 Antibody - C-terminal region (ARP49248_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49248_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf184. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C1orf184. Alternate names: NTM1B, C1orf184ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60048