Learn More
Invitrogen™ C1orf198 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552109
Description
Immunogen sequence: ALKSSQGSRS SSLDALGPTR KEEEASFWKI NAERSRGEGP EAEFQSLTPS QIKSMEKGEK VLPPCYRQEP APKDREAKVE RPSTLRQEQR PLPNVSTERE RPQPVQAFSS ALHEAAPSQL EGKLPSPDVR QDDGEDTLFS EPKFAQ Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 66%.
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma.
Specifications
C1orf198 | |
Polyclonal | |
Unconjugated | |
C1orf198 | |
C1orf198; chromosome 1 open reading frame 198; uncharacterized protein C1orf198 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84886 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.15 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H425 | |
C1orf198 | |
Recombinant protein corresponding to Human C1orf198. Recombinant protein control fragment (Product #RP-88755). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.