Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1orf201 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS15056R
Description
May positively contribute to the induction of apoptosis triggered by O(6)-methylguanine.Specifications
C1orf201 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
STPG1 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
90529 |
STPG1 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5TH74 | |
C1orf201; MAPO2; O(6)-methylguanine-induced apoptosis 2; O6-methylguanine-induced apoptosis 2; sperm tail PG-rich repeat containing 1; sperm-tail PG-rich repeat containing 1; Sperm-tail PG-rich repeat-containing protein 1; STPG1; UPF0490 protein C1orf201 | |
KLH conjugated synthetic peptide derived from human C1orf201. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title