Learn More
C1orf27 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 228441AP
Catalog No. 50-173-3954
Add to cart
Description
Description
May play a role in the trafficking of a subset of G-protein coupled receptors.
Specifications
Specifications
C1orf27
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4PJX1, Q5SWX8
C1orf27, hODR 4, LAG1 interacting protein, odr 4, ODR4, Protein odr 4 homolog, TTG1, TTG1A
Rabbit
IgG
150 μL
-20°C
Primary
226499, 54953
Western Blot
Unconjugated
ODR4
Liquid
ODR4
C1orf27 Fusion Protein Ag18802
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse