Learn More
C1orf31 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 242091AP
Catalog No. 50-173-4265
Add to cart
Description
Description
C1orf31, also known as COA6, is a conserved mitochondrial protein required for respiratory complex IV biogenesis. Recently study reported that C1orf31 is required for maintece of cytochrome c oxidase (CcO) subunit levels, and has role in the Cu delivery pathway to CcO. Defects in C1orf31 is associated with mitochondrial respiratory chain disease (MRCD). (24549041).
Specifications
Specifications
C1orf31
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5JTJ3, Q8BGD8
C1orf31, COA6
Rabbit
IgG
150 μL
-20°C
Primary
388753, 67892
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
COA6
Liquid
COA6
C1orf31 Fusion Protein Ag21525
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse