Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICB-1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551201

Catalog No. PA551201


Add to cart

Description

Description

The antibody detects endogenous levels of total C1orf38 / THEMIS2 protein.

May constitute a control point in macrophage inflammatory response, promoting LPS-induced TNF production.
Specifications

Specifications

ICB-1
Polyclonal
Unconjugated
THEMIS2
Liquid
THEMIS2
Fusion protein of human THEMIS2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.4 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q5TEJ8
C1orf38, ICB1
Rabbit
IgG
100 μL
-20°C
Primary
9473
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit