Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf43 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552733
Description
Immunogen sequence: AMKSRRGPHV PVGHNAPKDL KEEIDIRLSR VQDIKYEPQL LADDDARLLQ LETQGNQSCY NYLYRMKALD AIRTS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
General regulator of phagocytosis. Required to uptake Gram negative bacterium by macrophages. [UniProt].
Specifications
C1orf43 | |
Polyclonal | |
Unconjugated | |
C1orf43 | |
C1orf43; chromosome 1 open reading frame 43; HCV NS5A-transactivated protein 4; Hepatitis C virus NS5A-transactivated protein 4; HSPC012; NICE3; NICE-3; NS5ATP4; Protein NICE-3; S863-3; Uncharacterized protein C1orf43 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25912 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BWL3 | |
C1orf43 | |
Recombinant protein corresponding to Human C1orf43. Recombinant protein control fragment (Product #RP-90304). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction