Learn More
C1orf77 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 214561AP
Catalog No. 50-173-3427
Add to cart
Description
Description
The protein encoded by this gene is a protein serine/threonine phosphatase of the PP2C family. This protein can interact with integrin-linked kinase (ILK/ILK1), a regulator of integrin mediated signaling, and regulate the kinase activity of ILK. Through the interaction with ILK, this protein may selectively affect the signaling process of ILK-mediated glycogen synthase kinase 3 beta (GSK3beta), and thus participate in Wnt signaling pathway.
Specifications
Specifications
C1orf77
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9CY57, Q9Y3Y2
C1orf77, CHTOP, FOP, Friend of PRMT1 protein, pp7704, RP1 178F15.2
Rabbit
IgG
150 μL
-20°C
Primary
26097, 66511
Western Blot
Unconjugated
CHTOP
Liquid
Chtop
C1orf77 Fusion Protein Ag14736
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse