Learn More
C1q R1/CD93 Mouse anti-Human, DyLight 488, Clone: 2F7D11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237310G
Catalog No. NBP237310G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
C1q R1/CD93
2F7D11
Western Blot, ELISA, Immunohistochemistry
Purified
CD93
Purified recombinant fragment of human CD93 (AA: 474-535) expressed in E. Coli.
Protein A or G purified
B Cell Development and Differentiation Markers
Primary
22918
ELISA, Immunohistochemistry, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human