Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1QB Antibody (C1QB/2965) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279921
Description
C1QB Monoclonal specifically detects C1QB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
C1QB | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
C1QB/2965 | |
Unconjugated | |
10 mM PBS with No Preservative | |
C1QB | |
Recombinant fragment (around aa 41-188) of human C1QB protein (exact sequence is proprietary) (Uniprot: P02746) | |
100 μg | |
Primary | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction